KEY023878535 Key Organics / BIONET

(3-phenyl-1,2-oxazol-5-yl)methanamine

  • MDL number MFCD03844852
  • Cas Number : 54408-35-4
  • Molecular Weight : 174.203

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51 - 53
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82908

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6M-375S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart