KEY023878537 Key Organics / BIONET

(6E)-6-[(dimethylamino)methylidene]-1,3-dimethyl-1H,2H,3H,4H,5H,6H-furo[2,3-d]pyrimidine-2,4,5-trione

  • MDL number MFCD06200980
  • Cas Number : 453557-63-6
  • Molecular Weight : 251.242

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 248 - 250
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84043

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6T-0305_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart