KEY023878546 Key Organics / BIONET

2-{[4-(2,5-dimethyl-1H-pyrrol-1-yl)phenyl]methylidene}propanedinitrile

  • MDL number MFCD05663794
  • Cas Number : 771567-64-7
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 124 - 127
Iupac Name 2-[4-(2,5-dimethyl-1H-pyrrol-1-yl)benzylidene]malononitrile
Inchi Code 1S/C16H13N3/c1-12-3-4-13(2)19(12)16-7-5-14(6-8-16)9-15(10-17)11-18/h3-9H,1-2H3
Inchi Key SQSNNYNMFMHDSC-UHFFFAOYSA-N

Description

Melting Point 124 - 127
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84347

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6X-0707_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart