KEY023878547 Key Organics / BIONET

2-{[4-(2,5-dimethyl-1H-pyrrol-1-yl)phenyl]methyl}propanedinitrile

  • MDL number MFCD05663795
  • Cas Number : 771552-11-5
  • Molecular Weight : 249.317

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121 - 124
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84348

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6X-0708_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart