KEY023878548 Key Organics / BIONET

ethyl 1-(4-acetyl-2-nitrophenyl)piperidine-4-carboxylate

  • MDL number MFCD00205978
  • Cas Number : 217489-84-4
  • Molecular Weight : 320.345

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84349

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6X-0709_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart