KEY023878549 Key Organics / BIONET

1-[3-nitro-4-(piperidin-1-yl)phenyl]ethan-1-one

  • MDL number MFCD00047468
  • Cas Number : 30877-80-6
  • Molecular Weight : 248.282

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84350

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6X-0710_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart