KEY023878552 Key Organics / BIONET

methyl 6-{[(tert-butoxy)carbonyl]amino}pyridine-3-carboxylate

  • MDL number MFCD05663797
  • Cas Number : 144186-11-8
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 166 - 170
Iupac Name methyl 6-[(tert-butoxycarbonyl)amino]nicotinate
Inchi Code 1S/C12H16N2O4/c1-12(2,3)18-11(16)14-9-6-5-8(7-13-9)10(15)17-4/h5-7H,1-4H3,(H,13,14,16)
Inchi Key KHMRKCRGUHPQLA-UHFFFAOYSA-N

Description

Melting Point 166 - 170
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84353

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6X-0713_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart