KEY023878557 Key Organics / BIONET

1-[(4-fluorophenyl)sulfanyl]-4-nitrobenzene

  • MDL number MFCD04117815
  • Cas Number : 2438-85-9
  • Molecular Weight : 249.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92 - 95
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85245

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7G-445S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart