KEY023878558 Key Organics / BIONET

1-[2,3-dichloro-4-(ethylsulfanyl)phenyl]ethan-1-one

  • MDL number MFCD05663800
  • Cas Number : 105917-69-9
  • Molecular Weight : 249.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55 - 56
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85674

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7J-335S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart