KEY023878560 Key Organics / BIONET

6-methyl-3-(thiophen-2-yl)-1,2,4-triazin-5-ol

  • MDL number MFCD04117817
  • Cas Number : 886360-71-0
  • Molecular Weight : 193.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 292 - 294
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86402

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7M-541S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart