KEY023878561 Key Organics / BIONET

3-(3-chlorophenyl)-6-methyl-1,2,4-triazin-5-ol

  • MDL number MFCD04117818
  • Cas Number : 886360-72-1
  • Molecular Weight : 221.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206 - 208
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86404

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7M-543S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart