KEY023878563 Key Organics / BIONET

methyl 3-(4-methylbenzenesulfonamido)thiophene-2-carboxylate

  • MDL number MFCD00178119
  • Cas Number : 79128-72-6
  • Molecular Weight : 311.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147 - 151
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86876

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7N-592S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart