KEY023878566 Key Organics / BIONET

2-[(carboxymethyl)amino]propanoic acid hydrochloride

  • MDL number MFCD03844854
  • Cas Number : 33012-75-8
  • Molecular Weight : 183.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238 - 240
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87794

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7T-0622_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart