KEY023878567 Key Organics / BIONET

methyl 5-nitro-2-(piperazin-1-yl)benzoate

  • MDL number MFCD03791233
  • Cas Number : 886360-73-2
  • Molecular Weight : 265.269

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113 - 115
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87851

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7W-0206_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart