KEY023878572 Key Organics / BIONET

ethyl 4-chloro-5-formyl-2-(phenylamino)thiophene-3-carboxylate

  • MDL number MFCD02181910
  • Cas Number : 78267-24-0
  • Molecular Weight : 309.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118 - 120
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87917

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7W-0320_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart