KEY023878574 Key Organics / BIONET

methyl 2-(4-benzylpiperazin-1-yl)-5-nitrobenzoate

  • MDL number MFCD02643034
  • Cas Number : 381195-04-6
  • Molecular Weight : 355.394

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113 - 115
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87927

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7W-0334_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart