KEY023878575 Key Organics / BIONET

2-[4-(ethoxycarbonyl)piperidin-1-yl]-5-nitrobenzoic acid

  • MDL number MFCD03791236
  • Cas Number : 78243-32-0
  • Molecular Weight : 322.317

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173 - 175
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87928

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7W-0335_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart