KEY023878579 Key Organics / BIONET

methyl 2-(3-formylphenoxy)-5-nitrobenzoate

  • MDL number MFCD03844856
  • Cas Number : 886360-75-4
  • Molecular Weight : 301.254

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103 - 105
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87943

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7W-0357_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart