KEY023878580 Key Organics / BIONET

ethyl 2-[(2-methylphenyl)amino]-4-oxo-4,5-dihydrothiophene-3-carboxylate

  • MDL number MFCD03791239
  • Cas Number : 886360-76-5
  • Molecular Weight : 277.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139 - 141
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87950

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7W-0368_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart