KEY023878586 Key Organics / BIONET

methyl 1-methyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD00052747
  • Cas Number : 37619-24-2
  • Molecular Weight : 139.154

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88099

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7X-0419_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart