KEY023878587 Key Organics / BIONET

methyl 4-formyl-1-methyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD01571196
  • Cas Number : 67858-47-3
  • Molecular Weight : 167.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 96 - 98
Iupac Name methyl 4-formyl-1-methyl-1H-pyrrole-2-carboxylate
Inchi Code 1S/C8H9NO3/c1-9-4-6(5-10)3-7(9)8(11)12-2/h3-5H,1-2H3
Inchi Key GXSZDESAIZEFGZ-UHFFFAOYSA-N

Description

Melting Point 96 - 98
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88100

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7X-0420_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart