KEY023878589 Key Organics / BIONET

1-[3-amino-4-(morpholin-4-yl)phenyl]ethan-1-one

  • MDL number MFCD00116066
  • Cas Number : 217489-78-6
  • Molecular Weight : 220.272

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162 - 164
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88102

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7X-0700_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart