KEY023878592 Key Organics / BIONET

4-(morpholine-4-carbonyl)aniline

  • MDL number MFCD00158788
  • Cas Number : 51207-86-4
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 132 - 135
Iupac Name 4-(4-morpholinylcarbonyl)aniline
Inchi Code 1S/C11H14N2O2/c12-10-3-1-9(2-4-10)11(14)13-5-7-15-8-6-13/h1-4H,5-8,12H2
Inchi Key WEHVQIQNGXWTME-UHFFFAOYSA-N

Description

Melting Point 132 - 135
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88105

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7X-0704_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart