KEY023878594 Key Organics / BIONET

methyl 5-bromo-1-benzothiophene-2-carboxylate

  • MDL number MFCD05662034
  • Cas Number : 7312-11-0
  • Molecular Weight : 271.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 118 - 119
Iupac Name methyl 5-bromo-1-benzothiophene-2-carboxylate
Inchi Code 1S/C10H7BrO2S/c1-13-10(12)9-5-6-4-7(11)2-3-8(6)14-9/h2-5H,1H3
Inchi Key XDYVZHUZZZKQOS-UHFFFAOYSA-N

Description

Melting Point 118 - 119
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88107

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7X-0709_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart