KEY023878597 Key Organics / BIONET

ethyl 2-[(1-formylnaphthalen-2-yl)oxy]acetate

  • MDL number MFCD03001652
  • Cas Number : 76322-09-3
  • Molecular Weight : 258.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 90 - 92
Iupac Name ethyl [(1-formyl-2-naphthyl)oxy]acetate
Inchi Code 1S/C15H14O4/c1-2-18-15(17)10-19-14-8-7-11-5-3-4-6-12(11)13(14)9-16/h3-9H,2,10H2,1H3
Inchi Key SZSNBKWESSADJV-UHFFFAOYSA-N

Description

Melting Point 90 - 92
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88110

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7X-0712_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart