KEY023878600 Key Organics / BIONET

5-bromo-1-[(3-fluorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylic acid

  • MDL number MFCD05663804
  • Cas Number : 886360-92-5
  • Molecular Weight : 326.121

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166 - 169
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88113

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7X-0719_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart