KEY023878607 Key Organics / BIONET

2-butyl-5-chloro-6-nitro-1,3-benzoxazole

  • MDL number MFCD05663808
  • Cas Number : 886360-94-7
  • Molecular Weight : 254.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 55 - 57
Iupac Name 2-butyl-5-chloro-6-nitro-1,3-benzoxazole
Inchi Code 1S/C11H11ClN2O3/c1-2-3-4-11-13-8-5-7(12)9(14(15)16)6-10(8)17-11/h5-6H,2-4H2,1H3
Inchi Key LQMKZPJGVVIHSZ-UHFFFAOYSA-N

Description

Melting Point 55 - 57
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88236

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7X-5092_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart