KEY023878609 Key Organics / BIONET

2-butyl-5-nitro-1,3-benzoxazole

  • MDL number MFCD05663810
  • Cas Number : 886360-98-1
  • Molecular Weight : 220.228

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45 - 47
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88238

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7X-5094_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart