KEY023878613 Key Organics / BIONET

ethyl 1-(2-cyanophenyl)piperidine-4-carboxylate

  • MDL number MFCD04117820
  • Cas Number : 357670-16-7
  • Molecular Weight : 258.321

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45 - 47
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90949

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8T-0284_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart