KEY023878615 Key Organics / BIONET

ethyl 2-(methylamino)-4-oxo-4,5-dihydrothiophene-3-carboxylate

  • MDL number MFCD03791241
  • Cas Number : 886360-78-7
  • Molecular Weight : 201.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163 - 166
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91040

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8W-0225_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart