KEY023878621 Key Organics / BIONET

1-tert-butyl-3-methyl-1H-pyrazol-5-amine

  • MDL number MFCD00122731
  • Cas Number : 141459-53-2
  • Molecular Weight : 153.229

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65 - 67
Purity 95
Storage temp. RT
Physical Form White to Dark Orange Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91066

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/8W-0280_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart