KEY023878624 Key Organics / BIONET

3-tert-butyl-1-phenyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD02646688
  • Cas Number : 93045-47-7
  • Molecular Weight : 244.294

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204 - 206
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91087

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8W-0331_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart