KEY023878640 Key Organics / BIONET

4-(2-carboxybenzoyl)-1-methyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD05663822
  • Cas Number : 886361-15-5
  • Molecular Weight : 273.244

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 221 - 222
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91237

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8X-0722_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart