KEY023878642 Key Organics / BIONET

4-(4-methylbenzoyl)-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD05663824
  • Cas Number : 886361-19-9
  • Molecular Weight : 229.235

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 247 - 249
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91239

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8X-0724_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart