KEY023878653 Key Organics / BIONET

methyl 2-(2-butyl-1,3-benzoxazol-5-yl)acetate

  • MDL number MFCD05663830
  • Cas Number : 886361-26-8
  • Molecular Weight : 247.294

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91337

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8X-5074_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart