KEY023878659 Key Organics / BIONET

4-[1-(benzenesulfonyl)-1H-pyrrol-3-yl]-4-oxobutanoic acid

  • MDL number MFCD03791247
  • Cas Number : 81454-02-6
  • Molecular Weight : 307.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124 - 127
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93521

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9R-0053_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart