KEY023878666 Key Organics / BIONET

4-(2,6-dimethylmorpholin-4-yl)aniline

  • MDL number MFCD00051574
  • Cas Number : 218930-10-0
  • Molecular Weight : 206.289

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60 - 63
Purity 95
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94035

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9W-0242_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart