KEY023878670 Key Organics / BIONET

2-(4-fluorobenzenesulfonamido)acetic acid

  • MDL number MFCD02091617
  • Cas Number : 13029-71-5
  • Molecular Weight : 233.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158 - 160
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94069

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9W-0322_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart