KEY023878671 Key Organics / BIONET

2-chloro-4-(2,5-dimethyl-1H-pyrrol-1-yl)phenol

  • MDL number MFCD03844860
  • Cas Number : 647841-63-2
  • Molecular Weight : 221.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89 - 91
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94070

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9W-0323_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart