KEY023878673 Key Organics / BIONET

5-[3-(trifluoromethyl)phenyl]-2H-1,2,3,4-tetrazole

  • MDL number MFCD00068098
  • Cas Number : 92712-48-6
  • Molecular Weight : 214.151

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159 - 162
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94075

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9W-0332_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart