KEY023878675 Key Organics / BIONET

methyl 3-formyl-1H-indole-2-carboxylate

  • MDL number MFCD03848053
  • Cas Number : 18450-26-5
  • Molecular Weight : 203.197

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206 - 208
Purity 95
Storage temp. Store under N2 at -18
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94091

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P411,P422
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9W-0364_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart