KEY023878688 Key Organics / BIONET

2-methoxyethyl 2-{5-[2-chloro-4-(trifluoromethyl)phenoxy]-2-nitrophenoxy}propanoate

  • MDL number MFCD02102176
  • Cas Number : 72082-45-2
  • Molecular Weight : 463.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/104932

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MS-5008_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart