KEY040895150 Key Organics / BIONET

(Z)-N'-hydroxy-4-(1,3-oxazol-5-yl)benzene-1-carboximidamide

  • MDL number MFCD08276945
  • Cas Number : 860649-01-0
  • Molecular Weight : 203.201

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203 - 205
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3558

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10X-0862_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart