KEY040897339 Key Organics / BIONET

3-methyl-1H-1,2,4-triazole

  • MDL number MFCD00233931
  • Molecular Weight : 83.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 94 - 96
Iupac Name 3-methyl-1H-1,2,4-triazole
Inchi Code 1S/C3H5N3/c1-3-4-2-5-6-3/h2H,1H3,(H,4,5,6)
Inchi Key PZKFSRWSQOQYNR-UHFFFAOYSA-N

Description

Melting Point 94 - 96
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/104329

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-2909_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart