KEY040898003 Key Organics / BIONET

4,5-dibromo-N-(3-hydroxypropyl)-1H-pyrrole-2-carboxamide

  • MDL number MFCD06200910
  • Cas Number : 186956-98-9
  • Molecular Weight : 325.988

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160 - 162
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3515

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10X-0706_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart