KEY040898006 Key Organics / BIONET

methyl 2-amino-5-(4-chlorophenyl)-1,3-thiazole-4-carboxylate

  • MDL number MFCD06200914
  • Cas Number : 127918-92-7
  • Molecular Weight : 268.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238 - 240
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3622

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10X-0997_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart