KEY040898010 Key Organics / BIONET

{3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazol-5-yl}methanol

  • MDL number MFCD06200920
  • Cas Number : 886361-32-6
  • Molecular Weight : 244.173

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66 - 67
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7275

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11X-0245_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart