KEY040898013 Key Organics / BIONET

methyl 2-amino-5-(2-fluorophenyl)-1,3-thiazole-4-carboxylate

  • MDL number MFCD06200924
  • Cas Number : 886361-38-2
  • Molecular Weight : 252.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7338

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11X-0951_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart