KEY040898020 Key Organics / BIONET

4'-nitro-[1,1'-biphenyl]-4-carbaldehyde

  • MDL number MFCD06200931
  • Cas Number : 98648-23-8
  • Molecular Weight : 227.219

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117 - 120
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66490

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1Y-0709_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart