KEY040898021 Key Organics / BIONET

1-[5-(4-fluorophenyl)thiophen-2-yl]ethan-1-one

  • MDL number MFCD06200932
  • Cas Number : 886361-46-2
  • Molecular Weight : 220.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119 - 120
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66492

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1Y-0712_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart